Anti-PITX2 Rabbit Recombinant Polyclonal Antibody (FITC)
New Product
Type:
Antigen: PITX2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77835-050
- 77835-049
- Antigen symbol:
- PITX2
- PITX2
- Antigen name:
- Pituitary homeobox 2 protein (40-154AA)
- Pituitary homeobox 2 protein (40-154AA)
- Antigen synonyms:
- All1 responsive gene 1| ALL1 responsive protein ARP1| ALL1-responsive protein ARP1| ARP 1| ARP1| Brx 1| Brx1| Homeobox protein PITX2| IDG 2| IDG2| IGDS 2| IGDS| IGDS2| IHG 2| IHG2| IRID 2| IRID2| MGC111022| MGC20144| Otlx 2| Otlx2| Paired like homeodomain transcription factor 2| Paired-like homeodomain transcription factor 2| Pituitary homeo box 2| Pituitary homeobox 2| PITX 2| pitx2| PITX2_HUMAN| PTX 2| PTX2| RGS| RIEG 1| RIEG| Rieg bicoid related homeobox transcription factor 1| RIEG bicoid related homeobox transcription factor| RIEG bicoid-related homeobox transcription factor| RIEG1| RS| Solurshin
- All1 responsive gene 1| ALL1 responsive protein ARP1| ALL1-responsive protein ARP1| ARP 1| ARP1| Brx 1| Brx1| Homeobox protein PITX2| IDG 2| IDG2| IGDS 2| IGDS| IGDS2| IHG 2| IHG2| IRID 2| IRID2| MGC111022| MGC20144| Otlx 2| Otlx2| Paired like homeodomain transcription factor 2| Paired-like homeodomain transcription factor 2| Pituitary homeo box 2| Pituitary homeobox 2| PITX 2| pitx2| PITX2_HUMAN| PTX 2| PTX2| RGS| RIEG 1| RIEG| Rieg bicoid related homeobox transcription factor 1| RIEG bicoid related homeobox transcription factor| RIEG bicoid-related homeobox transcription factor| RIEG1| RS| Solurshin
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q99697
- Q99697
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Frequently Bought Together