Anti-KRCC1 Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: IF: 1:50 to 1:200
Type:
Antigen: KRCC1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77837-346
- 77837-345
- Antigen symbol:
- KRCC1
- KRCC1
- Antigen name:
- Lysine-rich coiled-coil protein 1 protein (1-259AA)
- Lysine-rich coiled-coil protein 1 protein (1-259AA)
- Antigen synonyms:
- KRCC1|CHBP2|BM-003|BM-044Lysine-rich coiled-coil protein 1|Cryptogenic hepatitis-binding protein 2
- KRCC1|CHBP2|BM-003|BM-044Lysine-rich coiled-coil protein 1|Cryptogenic hepatitis-binding protein 2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NPI7
- Q9NPI7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together