Anti-ARHGEF39 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ARHGEF39
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77833-351
- 77833-350
- Antigen symbol:
- ARHGEF39
- ARHGEF39
- Antigen name:
- Rho guanine nucleotide exchange factor 39 protein (22-197AA)
- Rho guanine nucleotide exchange factor 39 protein (22-197AA)
- Antigen synonyms:
- ARG39_HUMAN|Arhgef39|C9orf100|Chromosome 9 open reading frame 100|FLJ14642|Hypothetical protein LOC84904|MGC44886|Rho guanine nucleotide exchange factor (GEF) 39|Rho guanine nucleotide exchange factor 39|RP11 331F9.7|RP11-331F9.7|Vav like protein C9orf100
- ARG39_HUMAN|Arhgef39|C9orf100|Chromosome 9 open reading frame 100|FLJ14642|Hypothetical protein LOC84904|MGC44886|Rho guanine nucleotide exchange factor (GEF) 39|Rho guanine nucleotide exchange factor 39|RP11 331F9.7|RP11-331F9.7|Vav like protein C9orf100
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8N4T4
- Q8N4T4
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together