Anti-GPC Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: GPC
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Lymphocytic choriomeningitis virus
Specifications
- Catalog No:
- 77817-138
- 77817-137
- Antigen symbol:
- GPC
- GPC
- Antigen name:
- Recombinant Lymphocytic choriomeningitis virus Pre-glycoprotein polyprotein GP complex protein (266-498AA)
- Recombinant Lymphocytic choriomeningitis virus Pre-glycoprotein polyprotein GP complex protein (266-498AA)
- Antigen synonyms:
- GPC|GP-C|Segment|SPre-glycoprotein polyprotein GP complex|Pre-GP-C) [Cleaved into: Stable signal peptide|SSP)|Glycoprotein G1|GP1)|Glycoprotein G2|GP2)]
- GPC|GP-C|Segment|SPre-glycoprotein polyprotein GP complex|Pre-GP-C) [Cleaved into: Stable signal peptide|SSP)|Glycoprotein G1|GP1)|Glycoprotein G2|GP2)]
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Lymphocytic choriomeningitis virus
- Lymphocytic choriomeningitis virus
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P09991
- P09991
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together