Anti-LYZL2 Rabbit Recombinant Antibody (HRP (Horseradish Peroxidase))
New Product
Type:
Antigen: LYZL2
Clonality: Recombinant
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77832-493
- 77832-494
- Antigen symbol:
- LYZL2
- LYZL2
- Antigen name:
- Lysozyme-like protein 2 protein (63-194AA)
- Lysozyme-like protein 2 protein (63-194AA)
- Antigen synonyms:
- LYZL2Lysozyme-like protein 2|Lysozyme-2|EC 3.2.1.17
- LYZL2Lysozyme-like protein 2|Lysozyme-2|EC 3.2.1.17
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q7Z4W2
- Q7Z4W2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together