Anti-TMX2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TMX2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77811-192
- 77803-192
- Antigen symbol:
- TMX2
- TMX2
- Antigen name:
- Thioredoxin-related transmembrane protein 2 protein (49-102AA)
- Thioredoxin-related transmembrane protein 2 protein (49-102AA)
- Antigen synonyms:
- Cell proliferation inducing gene 26 protein|Cell proliferation-inducing gene 26 protein|CGI 31|Growth inhibiting gene 11|PDIA12|PIG26|Protein disulfide isomerase family A|member 12|Thioredoxin domain containing protein 14|Thioredoxin domain-containing protein 14|Thioredoxin related transmembrane protein 2|Thioredoxin-related transmembrane protein 2|tmx2|TMX2_HUMAN|TXNDC14
- Cell proliferation inducing gene 26 protein|Cell proliferation-inducing gene 26 protein|CGI 31|Growth inhibiting gene 11|PDIA12|PIG26|Protein disulfide isomerase family A|member 12|Thioredoxin domain containing protein 14|Thioredoxin domain-containing protein 14|Thioredoxin related transmembrane protein 2|Thioredoxin-related transmembrane protein 2|tmx2|TMX2_HUMAN|TXNDC14
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y320
- Q9Y320
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together