Anti-EXTL3 Rabbit Recombinant Polyclonal Antibody
Catalog # 77806-460
Supplier: AFG Bioscience
New Product
Specifications
- Antigen name:Exostosin-like 3 protein (2-245AA)
- Antigen symbol:EXTL3
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:O43909
- Antigen synonyms:botv|DKFZp686C2342|Exostoses (multiple)-like 3|Exostoses-like 3|Exostosin-like 3|EXT-related protein 1|EXTL1L|EXTL3|EXTL3_HUMAN|EXTR1|Glucuronyl-galactosyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferas|Glucuronyl-galactosyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase|Hereditary multiple exostoses gene isolog|KIAA0519|Multiple exostosis-like protein 3|Putative tumor suppressor protein EXTL3|REG|Reg receptor|REGR|RPR
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on Blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: EXTL3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human