Anti-KYAT3 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: KYAT3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77829-525
- 77829-524
- Antigen symbol:
- KYAT3
- KYAT3
- Antigen name:
- Kynurenine--oxoglutarate transaminase 3 protein (1-454AA)
- Kynurenine--oxoglutarate transaminase 3 protein (1-454AA)
- Antigen synonyms:
- CCBL 2|Cysteine conjugate beta lyase 2|Cysteine S conjugate beta lyase 2|Cysteine-S-conjugate beta-lyase 2|DKFZp547N1117|DKFZp667D0223|KAT 3|KAT III|KAT3|KAT3_HUMAN|KATIII|kyat3|Kynurenine aminotransferase 3|Kynurenine aminotransferase III|Kynurenine oxoglutarate transaminase 3|Kynurenine oxoglutarate transaminase III|Kynurenine--glyoxylate transaminase|Kynurenine--oxoglutarate transaminase 3|Kynurenine--oxoglutarate transaminase III|MGC9398|RBM1|RBMX L1|RBMXL 1|RBMXL1|RNA binding motif protein X linked like 1|RP11 82K18.3|RP4 531M19.2
- CCBL 2|Cysteine conjugate beta lyase 2|Cysteine S conjugate beta lyase 2|Cysteine-S-conjugate beta-lyase 2|DKFZp547N1117|DKFZp667D0223|KAT 3|KAT III|KAT3|KAT3_HUMAN|KATIII|kyat3|Kynurenine aminotransferase 3|Kynurenine aminotransferase III|Kynurenine oxoglutarate transaminase 3|Kynurenine oxoglutarate transaminase III|Kynurenine--glyoxylate transaminase|Kynurenine--oxoglutarate transaminase 3|Kynurenine--oxoglutarate transaminase III|MGC9398|RBM1|RBMX L1|RBMXL 1|RBMXL1|RNA binding motif protein X linked like 1|RP11 82K18.3|RP4 531M19.2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q6YP21
- Q6YP21
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together