Anti-HLA-DRB4 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: HLA-DRB4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77807-105
- 77807-104
- Antigen symbol:
- HLA-DRB4
- HLA-DRB4
- Antigen name:
- HLA class II histocompatibility antigen, DR beta 4 chain protein (30-227AA)
- HLA class II histocompatibility antigen, DR beta 4 chain protein (30-227AA)
- Antigen synonyms:
- DR 4|DR beta 4 chain|DR4|DRB1 transplantation antigen |DRB4|DRB4_HUMAN|HLA class II histocompatibility antigen|HLA class II histocompatibility antigen DR beta 4 chain |HLA-DRB4|Human leucocyte antigen DRB4 |Leukocyte antigen |Major histocompatibility complex class II DR beta 4 |MHC class II antigen DRB4|MHC class II antigen HLA DR beta |MHC class2 antigen |MHC HLA DR-beta chain
- DR 4|DR beta 4 chain|DR4|DRB1 transplantation antigen |DRB4|DRB4_HUMAN|HLA class II histocompatibility antigen|HLA class II histocompatibility antigen DR beta 4 chain |HLA-DRB4|Human leucocyte antigen DRB4 |Leukocyte antigen |Major histocompatibility complex class II DR beta 4 |MHC class II antigen DRB4|MHC class II antigen HLA DR beta |MHC class2 antigen |MHC HLA DR-beta chain
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P13762
- P13762
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together