Anti-SNTB1 Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: IF: 1:50 to 1:200
Type:
Antigen: SNTB1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77824-363
- 77824-362
- Antigen symbol:
- SNTB1
- SNTB1
- Antigen name:
- Beta-1-syntrophin protein (51-111AA)
- Beta-1-syntrophin protein (51-111AA)
- Antigen synonyms:
- 59 DAP|59 kDa dystrophin associated protein A1 basic component 1|59 kDa dystrophin-associated protein A1 basic component 1|A1B|Beta 1 syntrophin|Beta-1-syntrophin|BSYN2|DAPA1B|Dystrophin associated protein A1 59kDa basic component 1|FLJ22442|MGC111389|SNT2|SNT2B1|SNTB 1|SNTB1_HUMAN|Syntrophin 2|Syntrophin beta 1|Syntrophin beta 1 (dystrophin associated protein A1 59kDa basic component 1)|Syntrophin-2|Tax interaction protein 43|TIP 43|TIP-43
- 59 DAP|59 kDa dystrophin associated protein A1 basic component 1|59 kDa dystrophin-associated protein A1 basic component 1|A1B|Beta 1 syntrophin|Beta-1-syntrophin|BSYN2|DAPA1B|Dystrophin associated protein A1 59kDa basic component 1|FLJ22442|MGC111389|SNT2|SNT2B1|SNTB 1|SNTB1_HUMAN|Syntrophin 2|Syntrophin beta 1|Syntrophin beta 1 (dystrophin associated protein A1 59kDa basic component 1)|Syntrophin-2|Tax interaction protein 43|TIP 43|TIP-43
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q13884
- Q13884
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together