Anti-POGZ Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: POGZ
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77794-670
- 77794-669
- Antigen symbol:
- POGZ
- POGZ
- Antigen name:
- Pogo transposable element with ZNF domain protein (1-300AA)
- Pogo transposable element with ZNF domain protein (1-300AA)
- Antigen synonyms:
- KIAA0461|MGC71543|Pogo transposable element with ZNF domain|POGZ|POGZ_HUMAN|putative protein product of Nbla00003|SUHW5|suppressor of hairy wing homolog|Suppressor of hairy wing homolog 5|Zinc finger protein 280E|Zinc finger protein 635|ZNF280E|ZNF635|ZNF635M
- KIAA0461|MGC71543|Pogo transposable element with ZNF domain|POGZ|POGZ_HUMAN|putative protein product of Nbla00003|SUHW5|suppressor of hairy wing homolog|Suppressor of hairy wing homolog 5|Zinc finger protein 280E|Zinc finger protein 635|ZNF280E|ZNF635|ZNF635M
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q7Z3K3
- Q7Z3K3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together