Anti-WWP1 Rabbit Polyclonal Antibody
77804-688
New Product
- Antigen name:WW domain-containing protein 1
- Antigen symbol:WWP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q9H0M0
- Antigen synonyms:WWP1|NEDD4-like E3 ubiquitin-protein ligase WWP1|EC 2.3.2.26|Atrophin-1-interacting protein 5|AIP5|HECT-type E3 ubiquitin transferase WWP1|TGIF-interacting ubiquitin ligase 1|Tiul1|WW domain-containing protein 1
- Storage buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: WWP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat