Anti-TRIM38 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TRIM38
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77811-552
- 77803-552
- Antigen symbol:
- TRIM38
- TRIM38
- Antigen name:
- E3 ubiquitin-protein ligase TRIM38 protein (45-259AA)
- E3 ubiquitin-protein ligase TRIM38 protein (45-259AA)
- Antigen synonyms:
- TRIM38|RNF15|RORET|E3 ubiquitin-protein ligase TRIM38|EC 2.3.2.27|RING finger protein 15|RING-type E3 ubiquitin transferase TRIM38|Tripartite motif-containing protein 38|Zinc finger protein RoRet
- TRIM38|RNF15|RORET|E3 ubiquitin-protein ligase TRIM38|EC 2.3.2.27|RING finger protein 15|RING-type E3 ubiquitin transferase TRIM38|Tripartite motif-containing protein 38|Zinc finger protein RoRet
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O00635
- O00635
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together