Anti-ABCA12 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IF: 1:50 to 1:200
Type:
Antigen: ABCA12
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77832-671
- 77832-672
- Antigen symbol:
- ABCA12
- ABCA12
- Antigen name:
- ATP-binding cassette sub-family A member 12 protein (45-158AA)
- ATP-binding cassette sub-family A member 12 protein (45-158AA)
- Antigen synonyms:
- ABC transporter A family member 12|ABC transporter ABCA.12|ABC12||ABCAC_HUMAN|At|ATH16|ATP binding cassette 12|ATP binding cassette sub family A (ABC1) member 12|ATP binding cassette sub family A member 12|ATP binding cassette transporter 12|ATP-binding cassette 12|ATP-binding cassette sub-family A member 12|ATP-binding cassette transporter 12|Ichthyosis congenita II lamellar ichthyosis B|ICR2B|LI2|Putative ABC2 homolog 16
- ABC transporter A family member 12|ABC transporter ABCA.12|ABC12||ABCAC_HUMAN|At|ATH16|ATP binding cassette 12|ATP binding cassette sub family A (ABC1) member 12|ATP binding cassette sub family A member 12|ATP binding cassette transporter 12|ATP-binding cassette 12|ATP-binding cassette sub-family A member 12|ATP-binding cassette transporter 12|Ichthyosis congenita II lamellar ichthyosis B|ICR2B|LI2|Putative ABC2 homolog 16
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q86UK0
- Q86UK0
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together