Anti-TSSK6 Rabbit Polyclonal Antibody
Catalog # 77809-393
Supplier: AFG Bioscience
New Product
Specifications
- Antigen name:Synthesized peptide derived from the Internal region of TSSK 6.
- Antigen symbol:TSSK6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Size:50 µg
- Form:Liquid
- Gene ID:Q9BXA6
- Antigen synonyms:TSSK6|SSTK|FKSG82|Testis-specific serine/threonine-protein kinase 6|TSK-6|TSSK-6|Testis-specific kinase 6|EC 2.7.11.1|Cancer/testis antigen 72|CT72|Serine/threonine-protein kinase SSTK|Small serine/threonine kinase
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC: 1:100 to 1:300, ELISA: 1:40000
Type:
Antigen: TSSK6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human,Mouse,Rat