Anti-TIGIT Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IF: 1:50 to 1:200
Type:
Antigen: TIGIT
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77826-521
- 77826-522
- Antigen symbol:
- TIGIT
- TIGIT
- Antigen name:
- T-cell immunoreceptor with Ig and ITIM domains protein (22-141AA)
- T-cell immunoreceptor with Ig and ITIM domains protein (22-141AA)
- Antigen synonyms:
- DKFZp667A205|ENSMUSG00000071552|FLJ39873|RGD1563191|T cell immunoreceptor with Ig and ITIM domains|TIGIT|TIGIT_HUMAN|V-set and immunoglobulin domain containing 9|V-set and immunoglobulin domain-containing protein 9|V-set and transmembrane domain containing 3|V-set and transmembrane domain-containing protein 3|VSIG9|VSTM3|Washington University cell adhesion molecule|WUCAM
- DKFZp667A205|ENSMUSG00000071552|FLJ39873|RGD1563191|T cell immunoreceptor with Ig and ITIM domains|TIGIT|TIGIT_HUMAN|V-set and immunoglobulin domain containing 9|V-set and immunoglobulin domain-containing protein 9|V-set and transmembrane domain containing 3|V-set and transmembrane domain-containing protein 3|VSIG9|VSTM3|Washington University cell adhesion molecule|WUCAM
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q495A1
- Q495A1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together