Anti-TRIM23 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB:1:1000 to 1:5000, IHC:1:20 to 1:200
Type:
Antigen: TRIM23
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77811-529
- 77803-529
- Antigen symbol:
- TRIM23
- TRIM23
- Antigen name:
- E3 ubiquitin-protein ligase TRIM23 protein (1-280AA)
- E3 ubiquitin-protein ligase TRIM23 protein (1-280AA)
- Antigen synonyms:
- ADP ribosylation factor domain protein 1|ADP-ribosylation factor domain-containing protein 1|ARF domain protein 1|ARFD1|E3 ubiquitin-protein ligase TRIM23|GTP binding protein ARD 1|GTP-binding protein ARD-1|N-acetyltransferase ARD1 human homolog of|N-acetyltransferase homolog of S. cerevisiae ARD1|RING finger protein 46|TRI23_HUMAN|TRIM23|Tripartite motif protein TRIM23|Tripartite motif-containing protein 23
- ADP ribosylation factor domain protein 1|ADP-ribosylation factor domain-containing protein 1|ARF domain protein 1|ARFD1|E3 ubiquitin-protein ligase TRIM23|GTP binding protein ARD 1|GTP-binding protein ARD-1|N-acetyltransferase ARD1 human homolog of|N-acetyltransferase homolog of S. cerevisiae ARD1|RING finger protein 46|TRI23_HUMAN|TRIM23|Tripartite motif protein TRIM23|Tripartite motif-containing protein 23
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P36406
- P36406
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together