Anti-VPS25 Rabbit Polyclonal Antibody
Catalog # 77804-406
Supplier: AFG Bioscience
New Product
Specifications
- Antigen name:VPS25_HUMAN
- Antigen symbol:VPS25
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q9BRG1
- Antigen synonyms:Dermal papilla derived protein 9| Dermal papilla-derived protein 9| DERP9| EAP20| ELL associated protein of 20 kDa| ELL-associated protein of 20 kDa| ESCRT-II complex subunit VPS25| FAP20| HGNC:28122| hVps25| MGC10540| Vacuolar protein sorting 25 homolog (S. cerevisiae)| Vacuolar protein sorting associated protein 25| Vacuolar protein-sorting-associated protein 25| VPS25| VPS25_HUMAN
- Storage buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: VPS25
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat