Anti-SLC2A4RG Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SLC2A4RG
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77799-995
- 77799-994
- Antigen symbol:
- SLC2A4RG
- SLC2A4RG
- Antigen name:
- SLC2A4 regulator protein (1-100AA)
- SLC2A4 regulator protein (1-100AA)
- Antigen synonyms:
- GEF|GLUT4 enhancer factor|HDBP 1|HDBP-1|HDBP1|Huntington disease gene regulatory region binding protein 1|Huntington disease gene regulatory region-binding protein 1|S2A4R_HUMAN|Si 1 2 19|Si 1 2|SLC2A4 regulator|SLC2A4RG|Zinc finger protein 2 5 Glut4 enhancer factor
- GEF|GLUT4 enhancer factor|HDBP 1|HDBP-1|HDBP1|Huntington disease gene regulatory region binding protein 1|Huntington disease gene regulatory region-binding protein 1|S2A4R_HUMAN|Si 1 2 19|Si 1 2|SLC2A4 regulator|SLC2A4RG|Zinc finger protein 2 5 Glut4 enhancer factor
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NR83
- Q9NR83
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together