Anti-RNF40 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: RNF40
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77797-567
- 77797-566
- Antigen symbol:
- RNF40
- RNF40
- Antigen name:
- E3 ubiquitin-protein ligase BRE1B protein (471-724AA)
- E3 ubiquitin-protein ligase BRE1B protein (471-724AA)
- Antigen synonyms:
- 95 kDa retinoblastoma associated protein|95 kDa retinoblastoma protein binding protein|95 kDa retinoblastoma-associated protein|BRE 1B|BRE1 B|BRE1 E3 ubiquitin ligase homolog B|BRE1-B|BRE1B|BRE1B_HUMAN|DKFZp686K191|E3 ubiquitin protein ligase BRE1B|E3 ubiquitin-protein ligase BRE1B|KIAA0661|MGC13051|Rb associated protein|RBP 95|RBP95|RING finger protein 40|Ring finger protein 40 E3 ubiquitin protein ligase|RNF 40|Rnf40|STARING
- 95 kDa retinoblastoma associated protein|95 kDa retinoblastoma protein binding protein|95 kDa retinoblastoma-associated protein|BRE 1B|BRE1 B|BRE1 E3 ubiquitin ligase homolog B|BRE1-B|BRE1B|BRE1B_HUMAN|DKFZp686K191|E3 ubiquitin protein ligase BRE1B|E3 ubiquitin-protein ligase BRE1B|KIAA0661|MGC13051|Rb associated protein|RBP 95|RBP95|RING finger protein 40|Ring finger protein 40 E3 ubiquitin protein ligase|RNF 40|Rnf40|STARING
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O75150
- O75150
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together