Order Entry
Canada
ContactUsLinkComponent
Anti-DCLRE1B Rabbit Recombinant Polyclonal Antibody
Anti-DCLRE1B Rabbit Recombinant Polyclonal Antibody
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-DCLRE1B Rabbit Recombinant Polyclonal Antibody
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77840-713
  • 77840-714
  • Antigen symbol:
  • DCLRE1B
  • DCLRE1B
  • Antigen name:
  • 5' exonuclease Apollo protein (345-493AA)
  • 5' exonuclease Apollo protein (345-493AA)
  • Antigen synonyms:
  • DCLRE1B| SNM1B5' exonuclease Apollo| EC 3.1.-.-| DNA cross-link repair 1B protein| SNM1 homolog B| SNMIB| hSNM1B
  • DCLRE1B| SNM1B5' exonuclease Apollo| EC 3.1.-.-| DNA cross-link repair 1B protein| SNM1 homolog B| SNMIB| hSNM1B
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q9H816
  • Q9H816
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping temperature:
  • Shipped on Blue ice
  • Shipped on Blue ice

Specifications

Frequently Bought Together