Anti-PEAK1 Rabbit Polyclonal Antibody
77809-271
New Product
- Antigen name:Synthesized peptide derived from the N-terminal region of SgK269
- Antigen symbol:PEAK1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q9H792
- Antigen synonyms:FLJ21140|FLJ34483|KIAA2002|NKF3 kinase family member|PEAK1_HUMAN|pseudopodium enriched atypical kinase|Pseudopodium-enriched atypical kinase 1|SGK269|Sugen kinase 269|Tyrosine-protein kinase SgK269
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: WB: 1:500 to 1:2000, ELISA: 1:5000
Type:
Antigen: PEAK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human