Anti-UGT1A6 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: UGT1A6
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77811-965
- 77803-965
- Antigen symbol:
- UGT1A6
- UGT1A6
- Antigen name:
- UDP-glucuronosyltransferase 1-6 protein (278-410AA)
- UDP-glucuronosyltransferase 1-6 protein (278-410AA)
- Antigen synonyms:
- GNT1|HLUGP|HLUGP1|Phenol metabolizing UDP glucuronosyltransferase|Phenol-metabolizing UDP-glucuronosyltransferase|UD16_HUMAN|UDP glucuronosyltransferase 1 family polypeptide A6|UDP glucuronosyltransferase 1A6|UDP glycosyltransferase 1 family polypeptide A6|UDP-glucuronosyltransferase 1-6|UDP-glucuronosyltransferase 1-F|UDP-glucuronosyltransferase 1A6|UDPGT|UDPGT 1-6|UGT 1F|UGT-1F|UGT1|UGT1 06|UGT1*6|UGT1-06|UGT1.6|UGT1A6S|UGT1F
- GNT1|HLUGP|HLUGP1|Phenol metabolizing UDP glucuronosyltransferase|Phenol-metabolizing UDP-glucuronosyltransferase|UD16_HUMAN|UDP glucuronosyltransferase 1 family polypeptide A6|UDP glucuronosyltransferase 1A6|UDP glycosyltransferase 1 family polypeptide A6|UDP-glucuronosyltransferase 1-6|UDP-glucuronosyltransferase 1-F|UDP-glucuronosyltransferase 1A6|UDPGT|UDPGT 1-6|UGT 1F|UGT-1F|UGT1|UGT1 06|UGT1*6|UGT1-06|UGT1.6|UGT1A6S|UGT1F
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P19224
- P19224
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together