Anti-MYSM1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MYSM1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77828-044
- 77828-045
- Antigen symbol:
- 2A-DUB;2ADUB;Histone H2A deubiquitinase;Histone H2A deubiquitinase MYSM1;KIAA1915;Myb like SWIRM and MPN domains 1;Myb like, SWIRM and MPN domain containing protein;Myb like, SWIRM and MPN domains 1;Myb-like;Myb-like, SWIRM and MPN domains 1;MYSM 1;mysm1;MYSM1 Myb-like, SWIRM and MPN domains 1;MYSM1_HUMAN;RP4-592A1.1;SWIRM and MPN domain-containing protein 1
- 2A-DUB;2ADUB;Histone H2A deubiquitinase;Histone H2A deubiquitinase MYSM1;KIAA1915;Myb like SWIRM and MPN domains 1;Myb like, SWIRM and MPN domain containing protein;Myb like, SWIRM and MPN domains 1;Myb-like;Myb-like, SWIRM and MPN domains 1;MYSM 1;mysm1;MYSM1 Myb-like, SWIRM and MPN domains 1;MYSM1_HUMAN;RP4-592A1.1;SWIRM and MPN domain-containing protein 1
- Antigen name:
- Histone H2A deubiquitinase MYSM1 protein (170-311AA)
- Histone H2A deubiquitinase MYSM1 protein (170-311AA)
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q5VVJ2
- Q5VVJ2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together