Anti-GRIK4 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: GRIK4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77823-740
- 77823-739
- Antigen symbol:
- GRIK4
- GRIK4
- Antigen name:
- Glutamate receptor ionotropic, kainate 4 protein (826-956AA)
- Glutamate receptor ionotropic, kainate 4 protein (826-956AA)
- Antigen synonyms:
- EAA 1|EAA1|Excitatory amino acid receptor 1|GluK4|Glutamate receptor|Glutamate receptor ionotropic kainate 4|Glutamate receptor ionotropic kainate 4 precursor|Glutamate receptor KA 1|Glutamate receptor KA 1precursor|Glutamate receptor KA-1|Glutamate receptor KA1|ionotropic kainate 4|GRIK 4|GRIK|Grik4|GRIK4_HUMAN|KA 1|KA1|OTTHUMP00000045951|OTTHUMP00000231881
- EAA 1|EAA1|Excitatory amino acid receptor 1|GluK4|Glutamate receptor|Glutamate receptor ionotropic kainate 4|Glutamate receptor ionotropic kainate 4 precursor|Glutamate receptor KA 1|Glutamate receptor KA 1precursor|Glutamate receptor KA-1|Glutamate receptor KA1|ionotropic kainate 4|GRIK 4|GRIK|Grik4|GRIK4_HUMAN|KA 1|KA1|OTTHUMP00000045951|OTTHUMP00000231881
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q16099
- Q16099
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together