Anti-TP63 Rabbit Polyclonal Antibody
77811-350
New Product
- Antigen name:Tumor protein p73-like
- Antigen symbol:TP63
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q9H3D4
- Antigen synonyms:AIS|Amplified in squamous cell carcinoma|B(p51A)|B(p51B)|Chronic ulcerative stomatitis protein|CUSP|DN p63 alpha 1|DNp63|EEC3|id:ibd3516|Keratinocyte transcription factor|Keratinocyte transcription factor KET|KET|LMS|MGC115972|MGC192897|NBP|OFC8|OTTHUMP00000209732|OTTHUMP00000209733|OTTHUMP00000209734|OTTHUMP00000209735|OTTHUMP00000209737|OTTHUMP00000209738|OTTHUMP00000209739|OTTHUMP00000209740|OTTHUMP00000209741|OTTHUMP00000209742|OTTHUMP00000209743|OTTHUMP00000209744|p40|p51|P51/P63|p53-related protein p63|p53CP|p63|P63_HUMAN|p73H|p73L|RHS|SHFM4|TAp63alpha|TP53CP|TP53L|TP63|TP73L|Transformation related protein 63|Transformation-related protein 63|Trp53rp1|Trp63|Tumor protein 63|Tumor protein p53-competing protein|Tumor protein p53-like|Tumor protein p63|Tumor protein p63 deltaN isoform delta|Tumor protein p73|Tumor protein p73-like
- Storage buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3.
- Storage temperature:Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: TP63
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat