Order Entry
Canada
ContactUsLinkComponent
Anti-crfb12 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-crfb12 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-crfb12 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77814-307
  • 77814-308
  • Antigen symbol:
  • crfb12
  • crfb12
  • Antigen name:
  • Zebrafish Cytokine receptor family member B12 protein (106-130AA)
  • Zebrafish Cytokine receptor family member B12 protein (106-130AA)
  • Antigen synonyms:
  • Cytokine receptor family member B12, crfb12
  • Cytokine receptor family member B12, crfb12
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Reactivity:
  • Zebrafish
  • Zebrafish
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q1RM05
  • Q1RM05
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together