Anti-P4HA2 Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:20 to 1:200, IP: 1:200 to 1:2000
Type:
Antigen: P4HA2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77793-080
- 77793-081
- Antigen symbol:
- P4HA2
- P4HA2
- Antigen name:
- Prolyl 4-hydroxylase subunit alpha-2 protein (236-535AA)
- Prolyl 4-hydroxylase subunit alpha-2 protein (236-535AA)
- Antigen synonyms:
- 2-oxoglutarate-4-dioxygenase subunit alpha-2|4 PH alpha 2|4-PH alpha-2|C-P4Halpha(II)|collagen prolyl 4-hydroxylase alpha(II)|P4HA2|P4HA2_HUMAN|procollagen proline, 2 oxoglutarate 4 dioxygenase (proline 4 hydroxylase), alpha polypeptide II|procollagen proline, 2 oxoglutarate 4 dioxygenase alpha subunit, isoform 2|Procollagen-proline|procollagen-proline,2-oxoglutarate-4-dioxygenase subunit alpha-2|Prolyl 4 hydroxylase alpha 2 subunit|Prolyl 4 hydroxylase subunit alpha 2 [Precursor]|Prolyl 4-hydroxylase subunit alpha-2|Prolyl 4-hydroxylase, alpha polypeptide II
- 2-oxoglutarate-4-dioxygenase subunit alpha-2|4 PH alpha 2|4-PH alpha-2|C-P4Halpha(II)|collagen prolyl 4-hydroxylase alpha(II)|P4HA2|P4HA2_HUMAN|procollagen proline, 2 oxoglutarate 4 dioxygenase (proline 4 hydroxylase), alpha polypeptide II|procollagen proline, 2 oxoglutarate 4 dioxygenase alpha subunit, isoform 2|Procollagen-proline|procollagen-proline,2-oxoglutarate-4-dioxygenase subunit alpha-2|Prolyl 4 hydroxylase alpha 2 subunit|Prolyl 4 hydroxylase subunit alpha 2 [Precursor]|Prolyl 4-hydroxylase subunit alpha-2|Prolyl 4-hydroxylase, alpha polypeptide II
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O15460
- O15460
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.02% Sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% Sodium azide, 50% glycerol, pH 7.3
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together