Anti-TAB1 Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:20 to 1:200
Type:
Antigen: TAB1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
- Catalog No:
- 77825-498
- 77825-499
- Antigen symbol:
- TAB1
- TAB1
- Antigen name:
- TGF-beta-activated kinase 1 and MAP3K7-binding protein 1 protein (205-504AA)
- TGF-beta-activated kinase 1 and MAP3K7-binding protein 1 protein (205-504AA)
- Antigen synonyms:
- 2310012M03Rik|3'-Tab1|MAP3K7IP 1|MAP3K7IP1|MGC57664|Mitogen activated protein kinase kinase kinase 7 interacting protein 1|Mitogen-activated protein kinase kinase kinase 7-interacting protein 1|TAB 1|TAB1|TAB1_HUMAN|TAK1 binding protein 1|TAK1-binding protein 1|TGF beta activated kinase 1 binding protein 1|TGF-beta activated kinase 1/MAP3K7 binding protein 1|TGF-beta-activated kinase 1 and MAP3K7-binding protein 1|TGF-beta-activated kinase 1-binding protein 1|Transforming growth factor beta activated kinase binding protein 1
- 2310012M03Rik|3'-Tab1|MAP3K7IP 1|MAP3K7IP1|MGC57664|Mitogen activated protein kinase kinase kinase 7 interacting protein 1|Mitogen-activated protein kinase kinase kinase 7-interacting protein 1|TAB 1|TAB1|TAB1_HUMAN|TAK1 binding protein 1|TAK1-binding protein 1|TGF beta activated kinase 1 binding protein 1|TGF-beta activated kinase 1/MAP3K7 binding protein 1|TGF-beta-activated kinase 1 and MAP3K7-binding protein 1|TGF-beta-activated kinase 1-binding protein 1|Transforming growth factor beta activated kinase binding protein 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q15750
- Q15750
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together