Anti-KANSL1 Rabbit Recombinant Polyclonal Antibody (FITC)
New Product
Type:
Antigen: KANSL1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77832-067
- 77832-068
- Antigen symbol:
- KANSL1
- KANSL1
- Antigen name:
- KAT8 regulatory NSL complex subunit 1 protein (815-1086AA)
- KAT8 regulatory NSL complex subunit 1 protein (815-1086AA)
- Antigen synonyms:
- CENP-36| centromere protein 36| DKFZp686P06109| DKFZp727C091| hMSL1v1| hypothetical protein LOC284058| K1267_HUMAN| KANSL1| KAT8 regulatory NSL complex subunit 1| KDVS| KIAA1267| male-specific lethal 1 homolog| MGC102843| MLL1/MLL complex subunit KIAA1267| MSL1 homolog 1| MSL1v1| Non specific lethal 1 homolog| NSL complex protein NSL1| NSL1
- CENP-36| centromere protein 36| DKFZp686P06109| DKFZp727C091| hMSL1v1| hypothetical protein LOC284058| K1267_HUMAN| KANSL1| KAT8 regulatory NSL complex subunit 1| KDVS| KIAA1267| male-specific lethal 1 homolog| MGC102843| MLL1/MLL complex subunit KIAA1267| MSL1 homolog 1| MSL1v1| Non specific lethal 1 homolog| NSL complex protein NSL1| NSL1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q7Z3B3
- Q7Z3B3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Frequently Bought Together