Anti-HOPX Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: HOPX
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77834-802
- 77834-801
- Antigen symbol:
- HOPX
- HOPX
- Antigen name:
- Homeodomain-only protein (1-73AA)
- Homeodomain-only protein (1-73AA)
- Antigen synonyms:
- CAMEO|heart odd homeobox 1 protein|HOD|Homeodomain-only protein|HOP|HOP homeobox|HOP_HUMAN|HOPX|LAGY|Lung cancer-associated Y protein|NECC1|Not expressed in choriocarcinoma clone 1|Not expressed in choriocarcinoma protein 1|OB1|odd homeobox 1 protein|Odd homeobox protein 1|OTTHUMP00000158970|SMAP31|TOTO
- CAMEO|heart odd homeobox 1 protein|HOD|Homeodomain-only protein|HOP|HOP homeobox|HOP_HUMAN|HOPX|LAGY|Lung cancer-associated Y protein|NECC1|Not expressed in choriocarcinoma clone 1|Not expressed in choriocarcinoma protein 1|OB1|odd homeobox 1 protein|Odd homeobox protein 1|OTTHUMP00000158970|SMAP31|TOTO
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BPY8
- Q9BPY8
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together