Anti-ST5 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ST5
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77801-173
- 77801-172
- Antigen symbol:
- ST5
- ST5
- Antigen name:
- Suppression of tumorigenicity 5 protein (3-280AA)
- Suppression of tumorigenicity 5 protein (3-280AA)
- Antigen synonyms:
- DENND2B|DENN domain containing protein 2B|DENN domain-containing protein 2B|DENN/MADD domain containing 2B |HeLa tumor suppression 1|HTS1|p126|ST 5|ST5|ST5_HUMAN|Suppression of tumorigenicity 5|Suppression of tumorigenicity 5 protein
- DENND2B|DENN domain containing protein 2B|DENN domain-containing protein 2B|DENN/MADD domain containing 2B |HeLa tumor suppression 1|HTS1|p126|ST 5|ST5|ST5_HUMAN|Suppression of tumorigenicity 5|Suppression of tumorigenicity 5 protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P78524
- P78524
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together