Anti-SENP6 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SENP6
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77837-037
- 77837-038
- Antigen symbol:
- SENP6
- SENP6
- Antigen name:
- Sentrin-specific protease 6 protein (1-270AA)
- Sentrin-specific protease 6 protein (1-270AA)
- Antigen synonyms:
- 2810017C20Rik|FLJ11355|FLJ11887|KIAA0389|KIAA0797|SENP 6|Senp6|SENP6_HUMAN|Sentrin specific protease 6|Sentrin-specific protease 6|Sentrin/SUMO specific protease SENP6|Sentrin/SUMO-specific protease SENP6|SSP 1|SSP1|SUMO 1 specific protease 1|SUMO 1 specific protease|SUMO-1-specific protease 1|SUMO1 specific protease 1|SUMO1 specific protease|SUMO1/sentrin specific peptidase 6|SUSP 1|SUSP1
- 2810017C20Rik|FLJ11355|FLJ11887|KIAA0389|KIAA0797|SENP 6|Senp6|SENP6_HUMAN|Sentrin specific protease 6|Sentrin-specific protease 6|Sentrin/SUMO specific protease SENP6|Sentrin/SUMO-specific protease SENP6|SSP 1|SSP1|SUMO 1 specific protease 1|SUMO 1 specific protease|SUMO-1-specific protease 1|SUMO1 specific protease 1|SUMO1 specific protease|SUMO1/sentrin specific peptidase 6|SUSP 1|SUSP1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9GZR1
- Q9GZR1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together