Anti-FSD1 Rabbit Recombinant Polyclonal Antibody (HRP)
New Product
Type:
Antigen: FSD1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77835-243
- 77835-244
- Antigen symbol:
- FSD1
- FSD1
- Antigen name:
- Fibronectin type III and SPRY domain-containing protein 1 protein (251-370AA)
- Fibronectin type III and SPRY domain-containing protein 1 protein (251-370AA)
- Antigen synonyms:
- Fibronectin type 3 and SPRY (spla|ryanodine) domain containing (with coiled-coil motif) 1|Fibronectin type 3 and SPRY domain containing 1|Fibronectin type III and SPRY domain containing 1|Fibronectin type III and SPRY domain-containing protein 1|Fibronectin type III and SPRY domains containing protein 1||_HUMAN|GLFND|MGC3213|MGC58341|Microtubule-associated protein GLFND|MID1-related protein 1|midline 1-related protein 1|MIR1|VLP27
- Fibronectin type 3 and SPRY (spla|ryanodine) domain containing (with coiled-coil motif) 1|Fibronectin type 3 and SPRY domain containing 1|Fibronectin type III and SPRY domain containing 1|Fibronectin type III and SPRY domain-containing protein 1|Fibronectin type III and SPRY domains containing protein 1||_HUMAN|GLFND|MGC3213|MGC58341|Microtubule-associated protein GLFND|MID1-related protein 1|midline 1-related protein 1|MIR1|VLP27
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BTV5
- Q9BTV5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Frequently Bought Together