Anti-PCK1 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PCK1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77793-410
- 77793-411
- Antigen symbol:
- PCK1
- PCK1
- Antigen name:
- Phosphoenolpyruvate carboxykinase, cytosolic [GTP] protein (1-622AA)
- Phosphoenolpyruvate carboxykinase, cytosolic [GTP] protein (1-622AA)
- Antigen synonyms:
- cytosolic [GTP]|GTP|PCK1|PCKGC_HUMAN|PEP carboxykinase|PEPCK-C|PEPCK1|PEPCKC|Phosphoenolpyruvate carboxykinase 1 (soluble)|Phosphoenolpyruvate carboxykinase 1|Phosphoenolpyruvate carboxykinase|Phosphoenolpyruvate carboxykinase, cytosolic [GTP]|Phosphoenolpyruvate carboxykinase, cytosolic|Phosphoenolpyruvate carboxylase|Phosphopyruvate carboxylase
- cytosolic [GTP]|GTP|PCK1|PCKGC_HUMAN|PEP carboxykinase|PEPCK-C|PEPCK1|PEPCKC|Phosphoenolpyruvate carboxykinase 1 (soluble)|Phosphoenolpyruvate carboxykinase 1|Phosphoenolpyruvate carboxykinase|Phosphoenolpyruvate carboxykinase, cytosolic [GTP]|Phosphoenolpyruvate carboxykinase, cytosolic|Phosphoenolpyruvate carboxylase|Phosphopyruvate carboxylase
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P35558
- P35558
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together