Anti-USP9X Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: USP9X
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77833-875
- 77833-874
- Antigen symbol:
- USP9X
- USP9X
- Antigen name:
- Probable ubiquitin carboxyl-terminal hydrolase FAF-X protein (2360-2484AA)
- Probable ubiquitin carboxyl-terminal hydrolase FAF-X protein (2360-2484AA)
- Antigen synonyms:
- Deubiquitinating enzyme FAF X|Deubiquitinating enzyme FAF-X|DFFRX|Drosophila fat facets related X linked|FAF|Fafl|Fam|Fat facets homolog|Fat facets in mammals|Fat facets protein related X linked|Fat facets protein related, X-linked|Fat facets protein-related|hFAM|MRX99|Probable ubiquitin carboxyl terminal hydrolase FAF X|Probable ubiquitin carboxyl-terminal hydrolase FAF-X|Ubiquitin carboxyl-terminal hydrolase FAM|Ubiquitin specific peptidase 9 X linked|Ubiquitin specific peptidase 9, X-linked|Ubiquitin specific processing protease FAF X|Ubiquitin specific protease 9 X chromosome|Ubiquitin thioesterase FAF X|Ubiquitin thiolesterase FAF X|Ubiquitin thiolesterase FAF-X|Ubiquitin-specific protease 9|Ubiquitin-specific-processing protease FAF-X|USP9 (gene name)|Usp9x|USP9X_HUMAN|Uubiquitin specific protease 9, X chromosome (fat facets like Drosophila)|X chromosome|X-linked
- Deubiquitinating enzyme FAF X|Deubiquitinating enzyme FAF-X|DFFRX|Drosophila fat facets related X linked|FAF|Fafl|Fam|Fat facets homolog|Fat facets in mammals|Fat facets protein related X linked|Fat facets protein related, X-linked|Fat facets protein-related|hFAM|MRX99|Probable ubiquitin carboxyl terminal hydrolase FAF X|Probable ubiquitin carboxyl-terminal hydrolase FAF-X|Ubiquitin carboxyl-terminal hydrolase FAM|Ubiquitin specific peptidase 9 X linked|Ubiquitin specific peptidase 9, X-linked|Ubiquitin specific processing protease FAF X|Ubiquitin specific protease 9 X chromosome|Ubiquitin thioesterase FAF X|Ubiquitin thiolesterase FAF X|Ubiquitin thiolesterase FAF-X|Ubiquitin-specific protease 9|Ubiquitin-specific-processing protease FAF-X|USP9 (gene name)|Usp9x|USP9X_HUMAN|Uubiquitin specific protease 9, X chromosome (fat facets like Drosophila)|X chromosome|X-linked
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q93008
- Q93008
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together