Anti-PPP2R3C Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, IHC: 1:50 to 1:200
Type:
Antigen: PPP2R3C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Catalog No:
- 77819-310
- 77819-311
- Antigen symbol:
- PPP2R3C
- PPP2R3C
- Antigen name:
- Fusion protein of PPP2R3C
- Fusion protein of PPP2R3C
- Antigen synonyms:
- C14orf10|Chromosome 14 open reading frame 10|G4-1|G5pr|Hypothetical protein FLJ20644|P2R3C_HUMAN|protein phosphatase 2 (formerly 2A), regulatory subunit B'', gamma|Protein phosphatase 2 regulatory subunit B''gamma|protein phosphatase 2, regulatory subunit B'', gamma|Protein phosphatase subunit G5PR|rhabdomyosarcoma antigen MU-RMS-40.6A|Rhabdomyosarcoma antigen MU-RMS-40.6A/6C|rhabdomyosarcoma antigen Mu-RMS-40.6C|serine/threonine-protein phosphatase 2A regulatory subunit B'' subunit gamma
- C14orf10|Chromosome 14 open reading frame 10|G4-1|G5pr|Hypothetical protein FLJ20644|P2R3C_HUMAN|protein phosphatase 2 (formerly 2A), regulatory subunit B'', gamma|Protein phosphatase 2 regulatory subunit B''gamma|protein phosphatase 2, regulatory subunit B'', gamma|Protein phosphatase subunit G5PR|rhabdomyosarcoma antigen MU-RMS-40.6A|Rhabdomyosarcoma antigen MU-RMS-40.6A/6C|rhabdomyosarcoma antigen Mu-RMS-40.6C|serine/threonine-protein phosphatase 2A regulatory subunit B'' subunit gamma
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q969Q6
- Q969Q6
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together