Anti-FLT1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: FLT1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77810-754
- 77810-753
- Antigen symbol:
- FLT1
- FLT1
- Antigen name:
- Vascular endothelial growth factor receptor 1 protein (1048-1328AA)
- Vascular endothelial growth factor receptor 1 protein (1048-1328AA)
- Antigen synonyms:
- EC 2.7.10.1|FLT 1|FLT|Flt-1|FLT1|Fms like tyrosine kinase 1|Fms related tyrosine kinase 1|Fms related tyrosine kinase 1 (vascular endothelial growth factor/vascular permeability factor receptor)|Fms related tyrosine kinase 1 vascular endothelial growth factor/vascular permeability factor receptor|Fms-like tyrosine kinase 1|FRT|Soluble VEGF receptor 1 14|Soluble VEGFR1 variant 2|Soluble VEGFR1 variant 21|Tyrosine protein kinase FRT|Tyrosine protein kinase receptor FLT|Tyrosine-protein kinase FRT|Tyrosine-protein kinase receptor FLT|Vascular endothelial growth factor receptor 1|Vascular endothelial growth factor vascular permeability factor receptor|Vascular permeability factor receptor 1|Vascular permeability factor receptor|VEGFR 1|VEGFR-1|VEGFR1|VGFR1_HUMAN
- EC 2.7.10.1|FLT 1|FLT|Flt-1|FLT1|Fms like tyrosine kinase 1|Fms related tyrosine kinase 1|Fms related tyrosine kinase 1 (vascular endothelial growth factor/vascular permeability factor receptor)|Fms related tyrosine kinase 1 vascular endothelial growth factor/vascular permeability factor receptor|Fms-like tyrosine kinase 1|FRT|Soluble VEGF receptor 1 14|Soluble VEGFR1 variant 2|Soluble VEGFR1 variant 21|Tyrosine protein kinase FRT|Tyrosine protein kinase receptor FLT|Tyrosine-protein kinase FRT|Tyrosine-protein kinase receptor FLT|Vascular endothelial growth factor receptor 1|Vascular endothelial growth factor vascular permeability factor receptor|Vascular permeability factor receptor 1|Vascular permeability factor receptor|VEGFR 1|VEGFR-1|VEGFR1|VGFR1_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P17948
- P17948
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together