Anti-SEL1L Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SEL1L
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77836-732
- 77836-733
- Antigen symbol:
- SEL1L
- SEL1L
- Antigen name:
- Protein sel-1 homolog 1 protein (22-181AA)
- Protein sel-1 homolog 1 protein (22-181AA)
- Antigen synonyms:
- IBD2|PRO1063|Protein sel-1 homolog 1|SE1L1_HUMAN|Sel 1 like protein|Sel 1 suppressor of lin 12 like|Sel-1L|SEL1 LIKE|Sel1l|IKE|Suppressor of lin-12-like protein 1|TSA305|UNQ128|UNQ128/PRO1063
- IBD2|PRO1063|Protein sel-1 homolog 1|SE1L1_HUMAN|Sel 1 like protein|Sel 1 suppressor of lin 12 like|Sel-1L|SEL1 LIKE|Sel1l|IKE|Suppressor of lin-12-like protein 1|TSA305|UNQ128|UNQ128/PRO1063
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UBV2
- Q9UBV2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together