Anti-TM7SF2 Rabbit Polyclonal Antibody
New Product
Recommended dilution: ELISA: 1:2000 to 1:5000, WB: 1:500 to 1:2000, IHC: 1:25 to 1:100
Type:
Antigen: TM7SF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
- Catalog No:
- 77813-457
- 77813-456
- Antigen symbol:
- TM7SF2
- TM7SF2
- Antigen name:
- Synthetic peptide of TM7SF2
- Synthetic peptide of TM7SF2
- Antigen synonyms:
- TM7SF2|ANG1Delta(14)-sterol reductase TM7SF2|Delta-14-SR|EC 1.3.1.70|3-beta-hydroxysterol Delta|14)-reductase|Another new gene 1 protein|C-14 sterol reductase|C14SR|Putative sterol reductase SR-1|Sterol C14-reductase|Transmembrane 7 superfamily member 2
- TM7SF2|ANG1Delta(14)-sterol reductase TM7SF2|Delta-14-SR|EC 1.3.1.70|3-beta-hydroxysterol Delta|14)-reductase|Another new gene 1 protein|C-14 sterol reductase|C14SR|Putative sterol reductase SR-1|Sterol C14-reductase|Transmembrane 7 superfamily member 2
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O76062
- O76062
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together