Anti-SEPT3 Rabbit Polyclonal Antibody
Catalog # 77821-204
Supplier: AFG Bioscience
New Product
Specifications
- Antigen name:Synthesized peptide derived from internal of SEPT3
- Antigen symbol:SEPT3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q9UH03
- Antigen synonyms:bK250D10.3|MGC133218|Neuronal specific septin 3|Neuronal-specific septin-3|OTTHUMP00000028742|SEP 3|SEP3|SEPT 3|SEPT3 protein|SEPT3_HUMAN|Septin3
- Storage buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:3000
Type:
Antigen: SEPT3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse