Anti-TK1 Rabbit Recombinant Polyclonal Antibody
Catalog # 77802-643
Supplier: AFG Bioscience
New Product
Specifications
- Antigen name:Thymidine kinase, cytosolic protein (2-234AA)
- Antigen symbol:TK1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P04183
- Antigen synonyms:cytosolic|KITH_HUMAN|Thymidine kinase 1|Thymidine kinase 1 soluble|Thymidine kinase 1 soluble isoform|Thymidine kinase|Thymidine kinase cytosolic|TK 1|TK 2|TK1|Tk1a|Tk1b|TK2
- Storage buffer:0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:20 to 1:200
Type:
Antigen: TK1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human