Anti-SLC52A3 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SLC52A3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77838-664
- 77838-663
- Antigen symbol:
- SLC52A3
- SLC52A3
- Antigen name:
- Solute carrier family 52, riboflavin transporter, member 3 protein (159-220AA)
- Solute carrier family 52, riboflavin transporter, member 3 protein (159-220AA)
- Antigen synonyms:
- bA371L19.1|BVVLS|BVVLS1|C20orf54|C20orf54provided by HGNC|Chromosome 20 open reading frame 54|hRFT2|member 3|MGC10698|RFT2|RFVT3|Riboflavin transporter 2|riboflavin transporter|riboflavin transporter, member 3|S52A3_HUMAN|Slc52a3|solute carrier family 52 (riboflavin transporter), member 3|Solute carrier family 52|solute carrier family 52, riboflavin transporter, member 3
- bA371L19.1|BVVLS|BVVLS1|C20orf54|C20orf54provided by HGNC|Chromosome 20 open reading frame 54|hRFT2|member 3|MGC10698|RFT2|RFVT3|Riboflavin transporter 2|riboflavin transporter|riboflavin transporter, member 3|S52A3_HUMAN|Slc52a3|solute carrier family 52 (riboflavin transporter), member 3|Solute carrier family 52|solute carrier family 52, riboflavin transporter, member 3
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NQ40
- Q9NQ40
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together