Anti-Cytotoxin 4 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: Cytotoxin 4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Naja atra
Specifications
- Catalog No:
- 77816-829
- 77816-830
- Antigen symbol:
- Cytotoxin 4
- Cytotoxin 4
- Antigen name:
- Recombinant Naja atra Cytotoxin 4 protein (22-81AA)
- Recombinant Naja atra Cytotoxin 4 protein (22-81AA)
- Antigen synonyms:
- antibody|Cytotoxin 4|CTX-4M|CTX4|Cardiotoxin A4|CTX A4|Cardiotoxin analog IV|CTX IV
- antibody|Cytotoxin 4|CTX-4M|CTX4|Cardiotoxin A4|CTX A4|Cardiotoxin analog IV|CTX IV
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Naja atra
- Naja atra
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P01443
- P01443
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together