Anti-CTNNBL1 Rabbit Polyclonal Antibody
New Product
Recommended dilution: ELISA: 1:2000 to 1:5000, IHC: 1:50 to 1:200
Type:
Antigen: CTNNBL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
- Catalog No:
- 77832-852
- 77832-851
- Antigen symbol:
- CTNNBL1
- CTNNBL1
- Antigen name:
- Fusion protein of CTNNBL1
- Fusion protein of CTNNBL1
- Antigen synonyms:
- 5730471K09Rik|Beta catenin like 1|Beta catenin like protein 1|Beta-catenin-like protein 1|C20orf33|Cadherin associated protein|Catenin beta like 1|Chromosome 20 open reading frame 3|CTBL1_HUMAN|CTNN protein|CTNNBL 1|Ctnnbl1|dJ633O20.1|FLJ21108|NAP|Nuclear associated protein|nuclear protein NAP|Nuclear-associated protein|NYD SP19|OTTMUSP00000016933|P14|P14L|PP8304|RP5 1118M15.1|Testis development protein NYD SP19|Testis development protein NYD-SP19
- 5730471K09Rik|Beta catenin like 1|Beta catenin like protein 1|Beta-catenin-like protein 1|C20orf33|Cadherin associated protein|Catenin beta like 1|Chromosome 20 open reading frame 3|CTBL1_HUMAN|CTNN protein|CTNNBL 1|Ctnnbl1|dJ633O20.1|FLJ21108|NAP|Nuclear associated protein|nuclear protein NAP|Nuclear-associated protein|NYD SP19|OTTMUSP00000016933|P14|P14L|PP8304|RP5 1118M15.1|Testis development protein NYD SP19|Testis development protein NYD-SP19
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8WYA6
- Q8WYA6
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together