Anti-UAP1 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: UAP1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77823-243
- 77823-242
- Antigen symbol:
- UAP1
- UAP1
- Antigen name:
- UDP-N-acetylhexosamine pyrophosphorylase protein (1-522AA)
- UDP-N-acetylhexosamine pyrophosphorylase protein (1-522AA)
- Antigen synonyms:
- AGX 1| AGX| AGX-1| AGX-2| AGX1| Antigen X| AntigenX| SPAG 2| SPAG2| Sperm associated antigen 2| Sperm-associated antigen 2| UAP 1| uap1| UAP1_HUMAN| UDP N acetylhexosamine pyrophosphorylase| UDP N acteylglucosamine pyrophosphorylase 1| UDP-N-acetylglucosamine pyrophosphorylase
- AGX 1| AGX| AGX-1| AGX-2| AGX1| Antigen X| AntigenX| SPAG 2| SPAG2| Sperm associated antigen 2| Sperm-associated antigen 2| UAP 1| uap1| UAP1_HUMAN| UDP N acetylhexosamine pyrophosphorylase| UDP N acteylglucosamine pyrophosphorylase 1| UDP-N-acetylglucosamine pyrophosphorylase
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q16222
- Q16222
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together