Anti-MECR Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB:1:500 to 1:5000, IHC:1:20 to 1:200
Type:
Antigen: MECR
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Rat
Specifications
- Catalog No:
- 77842-888
- 77842-887
- Antigen symbol:
- MECR
- MECR
- Antigen name:
- Trans-2-enoyl-CoA reductase, mitochondrial protein (211-373AA)
- Trans-2-enoyl-CoA reductase, mitochondrial protein (211-373AA)
- Antigen synonyms:
- AI195831|CGI 63|FASN2B|Homolog of yeast 2 enoyl thioester reductase|HsNrbf-1|HsNrbf1|mecr|MECR_HUMAN|Mitochondrial 2 enoyl thioester reductase|mitochondrial|NRBF 1|NRBF-1|NRBF1|Nuclear receptor binding factor 1|Nuclear receptor-binding factor 1|OTTMUSP00000009996|RP23-13A13.1|Trans 2 enoyl CoA reductase|Trans-2-enoyl-CoA reductase
- AI195831|CGI 63|FASN2B|Homolog of yeast 2 enoyl thioester reductase|HsNrbf-1|HsNrbf1|mecr|MECR_HUMAN|Mitochondrial 2 enoyl thioester reductase|mitochondrial|NRBF 1|NRBF-1|NRBF1|Nuclear receptor binding factor 1|Nuclear receptor-binding factor 1|OTTMUSP00000009996|RP23-13A13.1|Trans 2 enoyl CoA reductase|Trans-2-enoyl-CoA reductase
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat
- Human, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BV79
- Q9BV79
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together