Anti-SMC4 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: SMC4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77842-395
- 77842-394
- Antigen symbol:
- SMC4
- SMC4
- Antigen name:
- Structural maintenance of chromosomes protein 4 protein (406-501AA)
- Structural maintenance of chromosomes protein 4 protein (406-501AA)
- Antigen synonyms:
- CAP C|CAPC|Chromosome associated polypeptide C|Chromosome-associated polypeptide C|hCAP C|hCAP-C|SMC 4|SMC protein 4|SMC-4|SMC4|SMC4_HUMAN|SMC4L1|Structural maintenance of chromosomes 4|Structural maintenance of chromosomes 4 like 1|Structural maintenance of chromosomes protein 4|XCAP C homolog|XCAP-C homolog
- CAP C|CAPC|Chromosome associated polypeptide C|Chromosome-associated polypeptide C|hCAP C|hCAP-C|SMC 4|SMC protein 4|SMC-4|SMC4|SMC4_HUMAN|SMC4L1|Structural maintenance of chromosomes 4|Structural maintenance of chromosomes 4 like 1|Structural maintenance of chromosomes protein 4|XCAP C homolog|XCAP-C homolog
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NTJ3
- Q9NTJ3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together