Anti-CHMP2A Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
New Product
Type:
Antigen: CHMP2A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77807-777
- 77807-778
- Antigen symbol:
- CHMP2A
- CHMP2A
- Antigen name:
- Charged multivesicular body protein 2a protein (1-222AA)
- Charged multivesicular body protein 2a protein (1-222AA)
- Antigen synonyms:
- BC 2|BC2|Charged multivesicular body protein 2a|CHM2A_HUMAN|CHMP2|CHMP2a|Chromatin modifying protein 2a|Chromatin-modifying protein 2a|hVps2 1|hVps2-1|putative breast adenocarcinoma marker|Putative breast adenocarcinoma marker BC-2|Putative breast adenocarcinoma marker BC2|Vacuolar protein sorting associated protein 2 1|Vacuolar protein sorting-associated protein 2-1|Vps2 1|VPS2|Vps2-1|VPS2A
- BC 2|BC2|Charged multivesicular body protein 2a|CHM2A_HUMAN|CHMP2|CHMP2a|Chromatin modifying protein 2a|Chromatin-modifying protein 2a|hVps2 1|hVps2-1|putative breast adenocarcinoma marker|Putative breast adenocarcinoma marker BC-2|Putative breast adenocarcinoma marker BC2|Vacuolar protein sorting associated protein 2 1|Vacuolar protein sorting-associated protein 2-1|Vps2 1|VPS2|Vps2-1|VPS2A
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O43633
- O43633
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together