Anti-ZNF462 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
New Product
Type:
Antigen: ZNF462
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77805-488
- 77805-487
- Antigen symbol:
- ZNF462
- ZNF462
- Antigen name:
- Zinc finger protein 462 protein (2223-2506AA)
- Zinc finger protein 462 protein (2223-2506AA)
- Antigen synonyms:
- DKFZp686B2325|DKFZp762N2316|FLJ14960|FLJ45904|KIAA1803|RP11-508N12.1|Zinc finger protein 462|ZN462_HUMAN|ZNF462
- DKFZp686B2325|DKFZp762N2316|FLJ14960|FLJ45904|KIAA1803|RP11-508N12.1|Zinc finger protein 462|ZN462_HUMAN|ZNF462
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96JM2
- Q96JM2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together